Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2,2'-Bifuran]-5,5'-dicarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A1329985-100MG A1329985-1G A1329985-5G
76657-942EA 30 USD InStock
76657-942 76657-944 76657-946
[2,2'-Bifuran]-5,5'-dicarboxylic acid 97%
[2,2'-Bifuran]-5,5'-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₆O₆
MW: 222.1510 g/mol
MDL Number: MFCD32174043
CAS Number: 50738-83-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty