Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-4-methyl-3-aminopyridine 95%

Supplier: AMBEED, INC
Warning

A154944-100MG A154944-10G A154944-1G A154944-250MG A154944-25G A154944-5G
76647-064EA 9 USD InStock
76647-064 76647-066 76647-068 76647-070 76647-072 76647-074
5-Bromo-4-methyl-3-aminopyridine 95%
5-Bromo-4-methyl-3-aminopyridine
Precaución: for Research Use Only


Formula: C₆H₇BrN₂
MW: 187.037 g/mol
MDL Number: MFCD07439976
CAS Number: 850892-12-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty