Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-(methoxycarbonyl)pyridine 1-oxide 97%

Supplier: AMBEED, INC
Warning

A155931-100MG A155931-1G A155931-250MG A155931-5G
76647-096EA 8 USD InStock
76647-096 76647-098 76647-100 76647-102
5-Bromo-2-(methoxycarbonyl)pyridine 1-oxide 97%
5-Bromo-2-(methoxycarbonyl)pyridine 1-oxide
Precaución: for Research Use Only


Formula: C₇H₆BrNO₃
MDL Number: MFCD28962490
CAS Number: 959741-32-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty