Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(4-(2-(3-(Fluoromethyl)azetidin-1-yl)ethoxy)phenyl)-3-(3-hydroxyphenyl)-4-methyl-2H-chromen-6-ol 98%

Supplier: AMBEED, INC
Warning

A161836-10MG A161836-1MG A161836-5MG
76647-156EA 227 USD InStock
76647-156 76647-158 76647-160
(S)-2-(4-(2-(3-(Fluoromethyl)azetidin-1-yl)ethoxy)phenyl)-3-(3-hydroxyphenyl)-4-methyl-2H-chromen-6-ol 98%
(S)-2-(4-(2-(3-(Fluoromethyl)azetidin-1-yl)ethoxy)phenyl)-3-(3-hydroxyphenyl)-4-methyl-2H-chromen-6-ol
Precaución: for Research Use Only


Formula: C₂₈H₂₈FNO₄
MDL Number: MFCD31631581
CAS Number: 1642297-01-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty