Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(methylsulfonyl)pyridine 95%

Supplier: AMBEED, INC
Warning

A179103-100MG A179103-1G A179103-250MG A179103-5G
76647-272EA 55 USD InStock
76647-272 76647-274 76647-276 77748-170
2-Bromo-4-(methylsulfonyl)pyridine 95%
2-Bromo-4-(methylsulfonyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₆BrNO₂S
MW: 236.0863 g/mol
MDL Number: MFCD28402467
CAS Number: 1193244-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty