Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxyindole-3-acetic acid 95%

Supplier: AMBEED, INC
Warning

A180751-100MG A180751-1G A180751-250MG A180751-5G
76647-284EA 14 USD InStock
76647-284 76647-286 76647-288 76647-290
6-Methoxyindole-3-acetic acid 95%
6-Methoxyindole-3-acetic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₃
MDL Number: MFCD01548400
CAS Number: 103986-22-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty