Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl pent-4-ynoate 95%

Supplier: AMBEED, INC
Warning

A185740-100MG A185740-1G A185740-250MG A185740-5G
76647-322EA 12 USD InStock
76647-322 76647-324 76647-326 76647-328
tert-Butyl pent-4-ynoate 95%
tert-Butyl pent-4-ynoate
Precaución: for Research Use Only


Formula: C₉H₁₄O₂
MDL Number: MFCD28361964
CAS Number: 185986-76-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty