Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Methylpiperazin-1-yl)benzaldehyde 97%

Supplier: AMBEED, INC
Warning

A190881-10G A190881-1G A190881-5G A190881-25G
76647-392EA 68 USD InStock
76647-392 76647-394 76647-396 77748-219
4-(4-Methylpiperazin-1-yl)benzaldehyde 97%
4-(4-Methylpiperazin-1-yl)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₂H₁₆N₂O
MDL Number: MFCD00452233
CAS Number: 27913-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty