Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl azetidin-3-yl(methyl)carbamate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A193783-1G A193783-250MG A193783-5G
76647-398EA 75 USD InStock
76647-398 76647-400 77748-234
Benzyl azetidin-3-yl(methyl)carbamate hydrochloride 98%
Benzyl azetidin-3-yl(methyl)carbamate hydrochloride
Precaución: for Research Use Only


Formula: C₁₂H₁₇ClN₂O₂
MW: 256.7286 g/mol
MDL Number: MFCD23102979
CAS Number: 1822874-05-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty