Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-bromo-3-ethylbenzoate 95%

Supplier: AMBEED, INC
Warning

A196606-100MG A196606-1G A196606-250MG
76647-426EA 108 USD InStock
76647-426 76647-428 76647-430
Ethyl 4-bromo-3-ethylbenzoate 95%
Ethyl 4-bromo-3-ethylbenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃BrO₂
MDL Number: MFCD22490682
CAS Number: 1822660-95-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty