Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-isopropylpicolinate 95%

Supplier: AMBEED, INC
Warning

A199943-100MG A199943-250MG
76647-460EA 30 USD InStock
76647-460 76647-462
Ethyl 3-isopropylpicolinate 95%
Ethyl 3-isopropylpicolinate
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO₂
MDL Number: MFCD31698495
CAS Number: 2119467-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty