Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lys01 trihydrochloride 99+%

Supplier: AMBEED, INC
Warning

A209517-10MG A209517-1MG A209517-50MG A209517-5MG A209517-250MG
76647-506EA 20 USD InStock
76647-506 76647-508 76647-510 76647-512 77622-260
Lys01 trihydrochloride 99+%
Lys01 trihydrochloride
Precaución: for Research Use Only


Formula: C₂₃H₂₆Cl₅N₅
MDL Number: MFCD28963913
CAS Number: 1391426-24-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty