Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 6-aminopicolinate 95%

Supplier: AMBEED, INC
Warning

A212667-100MG A212667-1G A212667-250MG A212667-5G
76647-520EA 85 USD InStock
76647-520 76647-522 76647-524 76647-526
tert-Butyl 6-aminopicolinate 95%
tert-Butyl 6-aminopicolinate
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂O₂
MW: 194.2304 g/mol
MDL Number: MFCD24127898
CAS Number: 1499890-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty