Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-(2-(2-Methoxyethoxy)ethoxy)ethoxy)thiophene 97%

Supplier: AMBEED, INC
Warning

A224526-100MG A224526-1G A224526-250MG
76647-570EA 31 USD InStock
76647-570 76647-572 76647-574
3-(2-(2-(2-Methoxyethoxy)ethoxy)ethoxy)thiophene 97%
3-(2-(2-(2-Methoxyethoxy)ethoxy)ethoxy)thiophene
Precaución: for Research Use Only


Formula: C₁₁H₁₈O₄S
MDL Number: MFCD31715118
CAS Number: 282540-12-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty