Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trifluoromethyl 4-fluorobenzenesulfonate 98%

Supplier: AMBEED, INC
Danger

A1339101-1G A1339101-250MG
76657-976EA 38 USD InStock
76657-976 76657-978
Trifluoromethyl 4-fluorobenzenesulfonate 98%
Trifluoromethyl 4-fluorobenzenesulfonate
Precaución: for Research Use Only


Formula: C₇H₄F₄O₃S
MDL Number: MFCD33023613
CAS Number: 2070902-77-9
UN: 2927
ADR: 6.1,8,Ⅱ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty