Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl trans-3-aminocyclobutane-1-carboxylate 98%

Supplier: AMBEED, INC
Danger

A232294-250MG A232294-5G
76647-630EA 188 USD InStock
76647-630 77748-396
tert-Butyl trans-3-aminocyclobutane-1-carboxylate 98%
tert-Butyl trans-3-aminocyclobutane-1-carboxylate
Precaución: for Research Use Only


Formula: C₉H₁₇NO₂
MDL Number: MFCD16661182
CAS Number: 1192547-86-6
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty