Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CY-09 97%

Supplier: AMBEED, INC
Warning

A234660-10MG A234660-1MG A234660-25MG A234660-5MG
76647-656EA 27 USD InStock
76647-656 76647-658 76647-660 76647-662
CY-09 97%
CY-09
Precaución: for Research Use Only


Formula: C₁₉H₁₂F₃NO₃S₂
MW: 423.43 g/mol
MDL Number: MFCD31619349
CAS Number: 1073612-91-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty