Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Ethylcyclobutanamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A252867-100MG A252867-1G A252867-250MG
76647-764EA 34 USD InStock
76647-764 76647-766 76647-768
N-Ethylcyclobutanamine hydrochloride 95%
N-Ethylcyclobutanamine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄ClN
MDL Number: MFCD19686391
CAS Number: 1334146-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty