Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-fluoro-6-formylbenzoate 95%

Supplier: AMBEED, INC
Warning

A257312-100MG A257312-250MG A257312-5G
76647-814EA 101 USD InStock
76647-814 76647-816 77748-480
Methyl 2-fluoro-6-formylbenzoate 95%
Methyl 2-fluoro-6-formylbenzoate
Precaución: for Research Use Only


Formula: C₉H₇FO₃
MW: 182.15 g/mol
MDL Number: MFCD24687162
CAS Number: 1256593-43-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty