Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3H-pyrazol-3-amine 98%

Supplier: AMBEED, INC
Danger

A261982-10G A261982-5G A261982-100G A261982-25G
76647-846EA 10 USD InStock
76647-846 76647-848 77748-496 77748-497
3H-pyrazol-3-amine 98%
3H-pyrazol-3-amine
Precaución: for Research Use Only


Formula: C₃H₅N₃
MW: 83.0919 g/mol
MDL Number: MFCD08700632
CAS Number: 1225387-53-0
UN: 3267
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty