Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-amino-2-iminoacetate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A270633-100MG A270633-1G A270633-250MG
76647-898EA 15 USD InStock
76647-898 76647-900 76647-902
Ethyl 2-amino-2-iminoacetate hydrochloride 95%
Ethyl 2-amino-2-iminoacetate hydrochloride
Precaución: for Research Use Only


Formula: C₄H₉ClN₂O₂
MDL Number: MFCD10697937
CAS Number: 76029-62-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty