Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dibromo-2-oxopropanoic acid 95%

Supplier: AMBEED, INC
Warning

A277602-250MG A277602-5G
76647-926EA 31 USD InStock
76647-926 76647-928
3,3-Dibromo-2-oxopropanoic acid 95%
3,3-Dibromo-2-oxopropanoic acid
Precaución: for Research Use Only


Formula: C₃H₂Br₂O₃
MDL Number: MFCD01686782
CAS Number: 600-35-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty