Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methylpyrimidine-4,6-dicarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A290573-100MG A290573-250MG
76648-012EA 70 USD InStock
76648-012 76648-014
5-Methylpyrimidine-4,6-dicarboxylic acid 97%
5-Methylpyrimidine-4,6-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₇H₆N₂O₄
MW: 182.1300 g/mol
MDL Number: MFCD31617065
CAS Number: 2010958-16-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty