Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-oxo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)spiro[indoline-3,4'-piperidine]-1'-carboxylate 95%

Supplier: AMBEED, INC
Warning

A301072-1G A301072-250MG A301072-100MG
76648-082EA 502 USD InStock
76648-082 76648-084 77748-582
tert-Butyl 2-oxo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)spiro[indoline-3,4'-piperidine]-1'-carboxylate 95%
tert-Butyl 2-oxo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)spiro[indoline-3,4'-piperidine]-1'-carboxylate
Precaución: for Research Use Only


Formula: C₂₃H₃₃BN₂O₅
MDL Number: MFCD16618542
CAS Number: 1253288-12-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty