Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,4R)-tert-Butyl 4-hydroxy-2-methylpyrrolidine-1-carboxylate 95%

Supplier: AMBEED, INC
Warning

A311614-100MG A311614-250MG A311614-5G
76648-172EA 21 USD InStock
76648-172 76648-174 76648-176
(2S,4R)-tert-Butyl 4-hydroxy-2-methylpyrrolidine-1-carboxylate 95%
(2S,4R)-tert-Butyl 4-hydroxy-2-methylpyrrolidine-1-carboxylate
Precaución: For research use only.


Formula: C₁₀H₁₉NO₃
MDL Number: MFCD11111254
CAS Number: 114676-61-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty