Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-((2R,3S,4S,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl octadec-9-enoate 98%

Supplier: AMBEED, INC
Warning

A316040-100MG A316040-10MG A316040-1MG A316040-50MG A316040-5MG
76648-190EA 251 USD InStock
76648-190 76648-192 76648-194 76648-196 76648-198
(E)-((2R,3S,4S,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl octadec-9-enoate 98%
(E)-((2R,3S,4S,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl octadec-9-enoate
Precaución: for Research Use Only


Formula: C₂₇H₄₅N₃O₆
MDL Number: MFCD19443751
CAS Number: 188181-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty