Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (8-bromooctyl)carbamate 95%

Supplier: AMBEED, INC
Warning

A328285-100MG A328285-1G A328285-250MG A328285-25G
76648-298EA 20 USD InStock
76648-298 76648-300 76648-302 77748-643
tert-Butyl (8-bromooctyl)carbamate 95%
tert-Butyl (8-bromooctyl)carbamate
Precaución: for Research Use Only


Formula: C₁₃H₂₆BrNO₂
MW: 308.26 g/mol
MDL Number: MFCD24473139
CAS Number: 142356-35-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty