Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxy-4-(trifluoromethyl)nicotinic acid 95%

Supplier: AMBEED, INC
Warning

A340859-100MG A340859-250MG A340859-50MG A340859-5G A340859-1G A340859-10G
76648-390EA 31 USD InStock
76648-390 76648-392 76648-394 76648-396 77593-992 77748-669
6-Hydroxy-4-(trifluoromethyl)nicotinic acid 95%
6-Hydroxy-4-(trifluoromethyl)nicotinic acid
Precaución: for Research Use Only


Formula: C₇H₄F₃NO₃
MW: 207.107 g/mol
MDL Number: MFCD06245493
CAS Number: 849020-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty