Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl trans-(methoxymethylene)acetate 97%

Supplier: AMBEED, INC
Warning

A359084-100G A359084-25G A359084-500G A359084-5G
76648-552EA 25 USD InStock
76648-552 76648-554 76648-556 76648-558
Methyl trans-(methoxymethylene)acetate 97%
Methyl trans-(methoxymethylene)acetate
Precaución: for Research Use Only


Formula: C₅H₈O₃
MW: 116.1152 g/mol
MDL Number: MFCD00128206
CAS Number: 5788-17-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty