Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(3-(2-Oxo-1,2-dihydropyridine-3-carboxamido)propyl)-5-phenylisoxazole-3-carboxamide 97%

Supplier: AMBEED, INC
Warning

A385405-10MG A385405-1MG A385405-25MG A385405-5MG A385405-50MG
76648-648EA 59 USD InStock
76648-648 76648-650 76648-652 76648-654 77622-298
N-(3-(2-Oxo-1,2-dihydropyridine-3-carboxamido)propyl)-5-phenylisoxazole-3-carboxamide 97%
N-(3-(2-Oxo-1,2-dihydropyridine-3-carboxamido)propyl)-5-phenylisoxazole-3-carboxamide
Precaución: for Research Use Only


Formula: C₁₉H₁₈N₄O₄
MDL Number: MFCD31619239
CAS Number: 1883510-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty