Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oct-7-ynoic acid 97%

Supplier: AMBEED, INC
Warning

A393928-100MG A393928-1G A393928-250MG A393928-5G A393928-25G
76648-734EA 26 USD InStock
76648-734 76648-736 76648-738 77551-176 77748-791
Oct-7-ynoic acid 97%
Oct-7-ynoic acid
Precaución: for Research Use Only


Formula: C₈H₁₂O₂
MDL Number: MFCD19228078
CAS Number: 10297-09-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty