Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LF3 97%

Supplier: AMBEED, INC
Warning

A406788-100MG A406788-1MG A406788-25MG A406788-50MG A406788-5MG
76648-790EA 184 USD InStock
76648-790 76648-792 76648-794 76648-796 76648-798 76648-800
LF3 97%
LF3
Precaución: for Research Use Only


Formula: C₂₀H₂₄N₄O₂S₂
MDL Number: MFCD03352482
CAS Number: 664969-54-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty