Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 95%

Supplier: AMBEED, INC
Warning

A407028-100MG A407028-1G A407028-250MG A407028-5G
76648-802EA 47 USD InStock
76648-802 76648-804 76648-806 76648-808
2,5-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 95%
2,5-Dichloro-7H-pyrrolo[2,3-d]pyrimidine
Precaución: for Research Use Only


Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
MDL Number: MFCD30537006
CAS Number: 2347374-92-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty