Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-dichloronicotinaldehyde 97%

Supplier: AMBEED, INC
Warning

A430937-100MG A430937-1G A430937-250MG
76648-932EA 117 USD InStock
76648-932 76648-934 76648-936
2-Amino-4,6-dichloronicotinaldehyde 97%
2-Amino-4,6-dichloronicotinaldehyde
Precaución: for Research Use Only


Formula: C₆H₄Cl₂N₂O
MDL Number: MFCD31543323
CAS Number: 2386318-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty