Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 6-chloro-5-nitropicolinate 95%

Supplier: AMBEED, INC
Warning

A432971-100MG A432971-250MG A432971-5G
76648-954EA 45 USD InStock
76648-954 76648-956 76648-958
Ethyl 6-chloro-5-nitropicolinate 95%
Ethyl 6-chloro-5-nitropicolinate
Precaución: for Research Use Only


Formula: C₈H₇ClN₂O₄
MDL Number: MFCD18250618
CAS Number: 1260669-90-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty