Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitroimidazole 95%

Supplier: AMBEED, INC
Warning

A435722-100G A435722-25G A435722-500G A435722-5G
76648-968EA 13 USD InStock
76648-968 76648-970 76648-972 76648-974
4-Nitroimidazole 95%
4-Nitroimidazole
Precaución: for Research Use Only


Formula: C₃H₃N₃O₂
MDL Number: MFCD00005196
CAS Number: 3034-38-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty