Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,6-Dibromopyridin-2-yl)methanol 95%

Supplier: AMBEED, INC
Warning

A438578-100MG A438578-1G A438578-250MG
76648-994EA 7 USD InStock
76648-994 76648-996 76648-998
(3,6-Dibromopyridin-2-yl)methanol 95%
(3,6-Dibromopyridin-2-yl)methanol
Precaución: for Research Use Only


Formula: C₆H₅Br₂NO
MDL Number: MFCD16606961
CAS Number: 1227601-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty