Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylbenzofuro[2,3-b]pyridin-8-yl trifluoromethanesulfonate 98%

Supplier: AMBEED, INC
Warning

A440783-100MG A440783-1G A440783-250MG A440783-5G
76649-012EA 7 USD InStock
76649-012 76649-014 76649-016 76649-018
2-Methylbenzofuro[2,3-b]pyridin-8-yl trifluoromethanesulfonate 98%
2-Methylbenzofuro[2,3-b]pyridin-8-yl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₁₃H₈F₃NO₄S
MDL Number: MFCD32633462
CAS Number: 1609373-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty