Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BLU-667 (Pralsetinib) 99%

Supplier: AMBEED, INC
Warning

A468588-1MG A468588-25MG A468588-5MG A468588-100MG A468588-10MG
76649-156EA 31 USD InStock
76649-156 76649-158 76649-160 77622-326 77622-328
BLU-667 (Pralsetinib) 99%
BLU-667 (Pralsetinib)
Precaución: for Research Use Only


Formula: C₂₇H₃₂FN₉O₂
MDL Number: MFCD31810155
CAS Number: 2097132-94-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty