Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoropyridin-3-amine 98%

Supplier: AMBEED, INC
Warning

A468899-250MG A468899-1G
76649-162EA 11 USD InStock
76649-162 77748-965
2,6-Difluoropyridin-3-amine 98%
2,6-Difluoropyridin-3-amine
Precaución: for Research Use Only


Formula: C₅H₄F₂N₂
MDL Number: MFCD13185331
CAS Number: 108118-69-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty