Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl phenylglyoxylate 95%

Supplier: AMBEED, INC
Warning

A477707-100G A477707-25G A477707-500G A477707-5G
76649-244EA 178 USD InStock
76649-244 76649-246 76649-248 76649-250
Ethyl phenylglyoxylate 95%
Ethyl phenylglyoxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
MDL Number: MFCD00009120
CAS Number: 1603-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty