Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-{[(2R,3S)-2-(2,4-Difluorophenyl)-3-methyloxiran-2-yl]methyl}-1H-1,2,4-triazole 98%

Supplier: AMBEED, INC
Warning

A497120-1G A497120-25G
76649-392EA 38 USD InStock
76649-392 76649-394
1-{[(2R,3S)-2-(2,4-Difluorophenyl)-3-methyloxiran-2-yl]methyl}-1H-1,2,4-triazole 98%
1-{[(2R,3S)-2-(2,4-Difluorophenyl)-3-methyloxiran-2-yl]methyl}-1H-1,2,4-triazole
Precaución: for Research Use Only


Formula: C₁₂H₁₁F₂N₃O
MDL Number: MFCD09833867
CAS Number: 127000-90-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty