Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

13-(Benzyloxy)-13-oxotridecanoic acid 95%

Supplier: AMBEED, INC
Warning

A498063-100MG A498063-250MG A498063-1G
76649-404EA 50 USD InStock
76649-404 76649-406 77749-021
13-(Benzyloxy)-13-oxotridecanoic acid 95%
13-(Benzyloxy)-13-oxotridecanoic acid
Precaución: for Research Use Only


Formula: C₂₀H₃₀O₄
MDL Number: MFCD31698716
CAS Number: 261178-55-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty