Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,3R)-3-Aminocyclohexanol hydrochloride 95%

Supplier: AMBEED, INC
Warning

A500955-100MG A500955-250MG A500955-5G
76649-416EA 37 USD InStock
76649-416 76649-418 76649-420
(1S,3R)-3-Aminocyclohexanol hydrochloride 95%
(1S,3R)-3-Aminocyclohexanol hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄ClNO
MDL Number: MFCD26954735
CAS Number: 2331211-57-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty