Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Chloro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: AMBEED, INC
Warning

A509271-100MG A509271-1G A509271-250MG
76649-474EA 44 USD InStock
76649-474 76649-476 77622-346
2-(2-Chloro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2-Chloro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₃H₁₈BClO₂
MW: 252.5448 g/mol
MDL Number: MFCD18729906
CAS Number: 1386860-54-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty