Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CB1954 (Tretazicar) 98%

Supplier: AMBEED, INC
Warning

A509901-10MG A509901-1MG A509901-25MG A509901-50MG
76649-486EA 30 USD InStock
76649-486 76649-488 76649-490 76649-492
CB1954 (Tretazicar) 98%
CB1954 (Tretazicar)
Precaución: for Research Use Only


Formula: C₉H₈N₄O₅
MW: 252.18 g/mol
MDL Number: MFCD00869490
CAS Number: 21919-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty