Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-vinyl-3-ethylimidazolium hexafluorophosphate 99% stabilized

Supplier: AMBEED, INC
Warning

A514388-1G A514388-25G A514388-5G
76649-524EA 9 USD InStock
76649-524 76649-526 76649-528
1-vinyl-3-ethylimidazolium hexafluorophosphate 99% stabilized
1-vinyl-3-ethylimidazolium hexafluorophosphate
Precaución: for Research Use Only


Formula: C₇H₁₁F₆N₂P
MDL Number: MFCD30470191
CAS Number: 1034364-43-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty