Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 2,2'-thiodiacetate 95%

Supplier: AMBEED, INC
Warning

A515130-100G A515130-1G A515130-25G A515130-500G A515130-5G
76649-532EA 45 USD InStock
76649-532 76649-534 76649-536 76649-538 76649-540
Dimethyl 2,2'-thiodiacetate 95%
Dimethyl 2,2'-thiodiacetate
Precaución: for Research Use Only


Formula: C₆H₁₀O₄S
MDL Number: MFCD00234244
CAS Number: 16002-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty