Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Azabicyclo[2.2.2]octane-2-carboxylic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A520285-100MG A520285-1G A520285-250MG A520285-5G A520285-25G
76649-558EA 10 USD InStock
76649-558 76649-560 76649-562 76649-564 77749-083
1-Azabicyclo[2.2.2]octane-2-carboxylic acid hydrochloride 97%
1-Azabicyclo[2.2.2]octane-2-carboxylic acid hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₄ClNO₂
MDL Number: MFCD00168044
CAS Number: 52601-23-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty