Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-(4-fluorophenyl)thiazole-5-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A542073-1G A542073-250MG A542073-25G A542073-5G
76649-656EA 17 USD InStock
76649-656 76649-658 76649-660 76649-662
2-Chloro-4-(4-fluorophenyl)thiazole-5-carbonitrile 98%
2-Chloro-4-(4-fluorophenyl)thiazole-5-carbonitrile
Precaución: for Research Use Only


Formula: C₁₀H₄ClFN₂S
MW: 238.6686 g/mol
MDL Number: MFCD30803418
CAS Number: 1628265-17-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty